missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, KnockDown
Specifications
Specifications
| Antigen | p53 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, KnockDown |
| Classification | Monoclonal |
| Clone | TRP/817 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, Knockout Validated, Knockdown Validated |
| Formulation | 10 mM PBS |
| Gene Alias | Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?