missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Antibody (PAb242) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£160.00 - £341.00
Specifications
| Antigen | p53 |
|---|---|
| Clone | PAb242 |
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256700
|
Novus Biologicals
NBP2-53113 |
0.1 mg |
£341.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684968
|
Novus Biologicals
NBP2-53113-0.025mg |
0.025 mg |
£160.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p53 Monoclonal specifically detects p53 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| p53 | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
| TP53 | |
| IgG1 | |
| Protein A purified |
| PAb242 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| 7157 | |
| SV40-transformed cell line BALB/c SVA31 E7 | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title