missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Antibody (CL2199), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£197.00 - £460.00
Specifications
| Antigen | p53 |
|---|---|
| Clone | CL2199 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18454882
|
Novus Biologicals
NBP2-34495-25ul |
25 μL |
£197.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18790714
|
Novus Biologicals
NBP2-34495 |
£460.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
p53 Monoclonal specifically detects p53 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated, PCR.Specifications
| p53 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| P04637 | |
| 7157 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KKGEPHHELPPGSTKRALPNNTSSSPQPKKKPLDGEYFTLQIRGRERFEMFRELNEALELKDAQAGKEPGGSRAHSSHLKSKKGQSTSR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL2199 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
| TP53 | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title