missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P4HTM Polyclonal Antibody

Description
This antibody does not react with mouse samples. Suggested positive control: antigen standard for PH (transient overexpression lysate), HeLa and human glioblastoma cell lysates.
Prolyl hydroxylase 4 is a prolyl hydroxylase that modifies HIF-alpha. Classic prolyl hydroxylases are found in the endoplasmic reticulum and modify collagen, whereas HIF is an intracellular protein and the prolyl hydroxylase sites do not resemble those modifying collagen. HIF is a transcriptional complex that plays a critical role in oxygen homeostasis. Prolyl hydroxylase is an essential component of the pathway through which cells sense oxygen. In the presence of oxygen, prolyl hydroxylases convert specific prolyl residues in HIF-alpha to hydroxyproline, leading to HIF-alpha destruction. Low oxygen levels, sensed at the cellular level, cause the HIF conversion to be reduced so that HIF is stable and there is increased angiogenesis. Prolyl hydroxylase 4, specifically, catalyzes the posttranslational formation of 4-hydroxyproline in HIF alpha proteins. It may function as a cellular oxygen sensor and, under normoxic conditions, may targets HIF through the hydroxylation for proteasomal degradation via the von Hippel-Lindau ubiquitylation complex.
Specifications
Specifications
| Antigen | P4HTM |
| Applications | Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | P4HTM |
| Gene Accession No. | Q8BG58, Q9NXG6 |
| Gene Alias | 4933406E20Rik; AI853847; BB128974; EGLN4; FLJ20262; HIFPH4; HIF-PH4; HIF-prolyl hydroxylase 4; HPH-4; hypoxia inducible factor prolyl 4 hydroxylase; hypoxia-inducible factor prolyl 4-hydroxylase; hypoxia-inducible factor prolyl hydroxylase 4; P4H TM; P4H with transmembrane domain; P4HTM; P4H-TM; PH 4; PH4; PH-4; PHD4; proline 4-hydroxylase; prolyl 4-hydroxylase, transmembrane; prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum); Prolyl hydroxlase domain-containing 4; RGD1311848; Transmembrane prolyl 4-hydroxylase |
| Gene Symbols | P4HTM |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?