missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P4HA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23185-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.Specifications
| P4HA3 | |
| Polyclonal | |
| Unconjugated | |
| P4HA3 | |
| 4 PH alpha 3, P4HA3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283208, 320452 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6W3F0, Q7Z4N8 | |
| P4HA3 | |
| P4HA3 Fusion Protein Ag19716 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction