missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P4HA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99486-100ul
This item is not returnable.
View return policy
Description
P4HA2 Polyclonal antibody specifically detects P4HA2 in Human samples. It is validated for Western Blot
Specifications
| P4HA2 | |
| Polyclonal | |
| Western Blot 1:1000-1:5000 | |
| 2-oxoglutarate-4-dioxygenase subunit alpha-2, 4-PH alpha 2, 4-PH alpha-2, alpha polypeptide II, collagen prolyl 4-hydroxylase alpha(II), C-P4Halpha(II), EC 1.14.11.2, Procollagen-proline, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), prolyl 4-hydroxylase subunit alpha-2, prolyl 4-hydroxylase, alpha polypeptide II | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human P4HA2. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8974 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction