missing translation for 'onlineSavingsMsg'
Learn More

P4HA2 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18394673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394673 Supplier Novus Biologicals Supplier No. NBP306468R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

P4HA2 Polyclonal antibody specifically detects P4HA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen P4HA2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias 2-oxoglutarate-4-dioxygenase subunit alpha-2, 4-PH alpha 2, 4-PH alpha-2, alpha polypeptide II, collagen prolyl 4-hydroxylase alpha(II), C-P4Halpha(II), EC 1.14.11.2, Procollagen-proline, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), prolyl 4-hydroxylase subunit alpha-2, prolyl 4-hydroxylase, alpha polypeptide II
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human P4HA2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8974
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.