missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P4HA2 Antibody (CL0351), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52922
This item is not returnable.
View return policy
Description
P4HA2 Monoclonal antibody specifically detects P4HA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Specifications
| P4HA2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8974 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| CL0351 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated | |
| 2-oxoglutarate-4-dioxygenase subunit alpha-2, 4-PH alpha 2, 4-PH alpha-2, alpha polypeptide II, collagen prolyl 4-hydroxylase alpha(II), C-P4Halpha(II), EC 1.14.11.2, Procollagen-proline, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), prolyl 4-hydroxylase subunit alpha-2, prolyl 4-hydroxylase, alpha polypeptide II | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LKEYILVEEAKLSKIKSWANKMEALTSKSAADAEGYLAHPVNAYKLVKRLNTDWPALEDLVLQDSAAGFIANLSVQRQFFPTDEDEIGAAKALMRLQDTYRLDPGTISRGELPGTKYQAMLSVDDCFGM | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction