missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p47phox Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104250
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NCF1/p47-phox.
p47phox (NCF1) is required, along with NCF2 and membrane bound cytochrome b558, for activation of the latent NADPH oxidase. This oxidase is a multicomponent enzyme that is activated to produce superoxide anion. Mutations in the gene can result in granulomatous disease.
Specifications
| p47phox | |
| Polyclonal | |
| Unconjugated | |
| Ncf1 | |
| 47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; NADPH oxidase organizer 2; NADPH oxidase subunit (47 kD); NADPH oxidase subunit (47kDa); Ncf1; NCF-1; NCF1A; NCF-47K; neutrop; neutrophil cytosol factor 1; neutrophil cytosolic factor 1; neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1); neutrophil NADPH oxidase factor 1; Nox organizer 2; NOXO2; nox-organizing protein 2; P47 phox; p47>phox>; p47phox; p47-phox; phagocyte oxidase 47 kDa; SH3 and PX domain-containing protein 1A; SH3PXD1A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114553, 17969, 653361 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| F1M707, P14598, Q09014 | |
| Ncf1 | |
| A synthesized peptide derived from human NCF1(Accession P14598), corresponding to amino acid residues P346-V390. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction