missing translation for 'onlineSavingsMsg'
Learn More

p400 Antibody, Novus Biologicals™

Product Code. 18413579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413579 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413579 Supplier Novus Biologicals Supplier No. H00057634B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

p400 Polyclonal antibody specifically detects p400 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen p400
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. AAH37208.1
Gene Alias CAG repeat protein 32, CAGH32hDomino, DKFZP434I225, Domino homolog, E1A binding protein p400, E1A-binding protein p400, EC 3.6.1, EC 3.6.4.-, FLJ45115, KIAA1498p400 kDa SWI2/SNF2-related protein, KIAA1818, P400, p400 SWI2/SNF2-related protein, TNRC12FLJ42018, trinucleotide repeat containing 12, Trinucleotide repeat-containing gene 12 protein
Host Species Mouse
Immunogen EP400 (AAH37208.1, 1 a.a. - 985 a.a.) full-length human protein. MHHGTGPQNVQHQLQRSRACPGSEGEEQPAHPNPPPSPAAPFAPSASPSAPQSPSYQIQQLMNRSPATGQNVNITLQSVGPVVGGNQQITLAPLPLPSPTSPGFQFSAQPRRFEHGSPSYIQVTSPLSQQVQTQSPTQPSPGPGQALQNVRAGAPGPGLGLCSSSPTGGFVDASVLVRQISLSPSSGGHFVFQDGSGLTQIAQGAQVQLQHPGTPITVRERRPSQPHTQSGGTIHHLGPQSPAAAGGAGLQPLASPSHITTANLPPQISSIIQGQLVQQQQVLQGPPLPRPLGFERTPGVLLPGAGGAAGFGMTSPPPPTSPSRTAVPPGLSSLPLTSVGNTGMKKVPKKLEEIPPASPEMAQMRKQCLDYHYQEMQALKEVFKEYLIELFFLQHFQGNMMDFLAFKKKHYAPLQAYLRQNDLDIEEEEEEEEEEEEKSEVINDEVKVVTGKDGQTGTPVAIATQLPPKVSAAFSSQQQPFQQALAGSLVAGAGSTVETDLFKRQQAMPSTGMAEQSKRPRLEVGHQGVVFQHPGAGAGVPLQQLMPTAQGGMPPTPQAAQLAGQRQSQQQYDPSTGPPVQNAASLHTPLPQLPGRLPPAGVPTAALSSALQFAQQPQVVEAQTQLQIPVKTQQPNVPIPAPPSSQLPIPPSQPAQLALHVPTPGKVQVQASQLSSLPQMVASTRLPVDPAPPCPRPLPTSSTSSLAPVSGSGPGPSPARSSPVNRPSSATNKALSPVTSRTPGVVASAPTKPQSPAQNATSSQDSSQDTLTEQITLENQVHQRIAELRKAGLWSQRRLPKLQEAPRPKPHWDYLLEEMQWMATDFAQERRWKVAAAKKLVRTVVRHHEEKQLREERGKKEEQSRLRRIAASTAREIECFWSNIEQVVEIKLRVELEEKRKKALNLQKVSRRGKELRPKGFDALQESSLDSGMSGRKRKASISLTDDEVDDEEETIEEEEANEGVVDHQTELSNLAKEGRLLLPY
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 57634
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.