missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p400 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35225-20ul
This item is not returnable.
View return policy
Description
p400 Polyclonal antibody specifically detects p400 in Human samples. It is validated for ELISA,Western Blot
Specifications
| p400 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| CAG repeat protein 32, CAGH32hDomino, DKFZP434I225, Domino homolog, E1A binding protein p400, E1A-binding protein p400, EC 3.6.1, EC 3.6.4.-, FLJ45115, KIAA1498p400 kDa SWI2/SNF2-related protein, KIAA1818, P400, p400 SWI2/SNF2-related protein, TNRC12FLJ42018, trinucleotide repeat containing 12, Trinucleotide repeat-containing gene 12 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-330 of human p400 (NP_056224.3).,, Sequence:, TLAPLPLPSPTSPGFQFSAQPRRFEHGSPSYIQVTSPLSQQVQTQSPTQPSPGPGQALQNVRAGAPGPGLGLCSSSPTGGFVDASVLVRQISLSPSSGGHFVFQDGSGLTQIAQGAQVQLQHPGTPITVRERRPSQPHTQSGGTIHHLGPQSPAAAGGAGLQPLASPSHITTANLPPQISSIIQGQLVQQQQVLQGPPLPRPLGFERTPGVLLPGAGGAAGFGMTSPPPPTSPSRTAVPPG | |
| 20 μL | |
| Cancer, Tumor Suppressors | |
| 57634 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction