missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p40 Rabbit anti-Human, Mouse, Rat, Clone: 10E2A4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | p40 |
|---|---|
| Clone | 10E2A4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18335934
|
Novus Biologicals
NBP3-15815-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375293
|
Bio-Techne
NBP3-15815-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p40 Monoclonal antibody specifically detects p40 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| p40 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10244 | |
| IgG | |
| Affinity purified |
| 10E2A4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 40 kDa Rab9 effector protein, bA65N13.1, p40, Rab9 effector p40, Rab9 effector protein with kelch motifs, RAB9P40DKFZp686P1077, RABEPK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 273-372 of human RAB9p40 (Q7Z6M1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title