missing translation for 'onlineSavingsMsg'
Learn More

P3HT (OPV grade - 91-94% RR)

Product Code. p-7214313
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 17176574

Brand: Thermo Scientific Alfa Aesar H66726.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104934-50-1 | (C10H14S)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104934-50-1
Molecular Formula (C10H14S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00217686
InChI Key DUFPJSOXRHVDOV-UHFFFAOYSA-N
Synonym Poly[3-hexylthiophene-2,5-diyl]
IUPAC Name 3-hexyl-2,5-dimethylthiophene
SMILES CCCCCCC1=C(-*)SC(-*)=C1

Specifications

Quantity 500 mg
Physical Form Solid
Chemical Name or Material P3HT

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.