missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P3H3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16023-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the leprecan family of proteoglycans, which function as collagen prolyl hydroxylases that are required for proper collagen biosynthesis, folding and assembly. This protein, like other family members, is thought to reside in the endoplasmic reticulum. Epigenetic inactivation of this gene is associated with breast and other cancers, suggesting that it may function as a tumor suppressor.Specifications
| P3H3 | |
| Polyclonal | |
| Unconjugated | |
| P3H3 | |
| GRCB, HSU47926, leprecan like 2, Leprecan like protein 2, LEPREL2, P3H3, Prolyl 3 hydroxylase 3, Protein B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10536, 14789, 297595 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CG70, Q8IVL6 | |
| P3H3 | |
| P3H3 Fusion Protein Ag8850 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction