missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p38 MAPK Recombinant Rabbit Monoclonal Antibody (23GB4530), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | p38 MAPK |
|---|---|
| Clone | 23GB4530 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30087609
|
Invitrogen™
MA553758 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089566
|
Invitrogen™
MA553759 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p38 MAPK Recombinant Monoclonal antibody specifically detects p38 MAPK in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| p38 MAPK | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P47811, P70618, Q16539 | |
| 1432, 26416, 81649 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4530 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Crk1; CSAID-binding protein; Csaids binding protein; CSBP; CSBP1; CSBP2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Exip; Hog; MAP kinase 11; MAP kinase 13; MAP kinase 14; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38 delta; MAPK 11; MAPK 13; MAPK 14; Mapk11; Mapk13; MAPK-13; Mapk14; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 13; mitogen activated protein kinase 14; mitogen-activated protein kinase 11; Mitogen-activated protein kinase 13; mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; mitogen-activated protein kinase p38 alpha; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38 delta; mitogen-activated protein kinase p38-2; MXI2; p38; p38 alpha; p38 delta MAP kinase; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38-2; p38a; p38ALPHA; p38-alpha; p38alpha Exip; P38b; p38Beta; p38beta2; p38delta; p38Hog; p38MAPK; Prkm11; PRKM13; PRKM14; Prkm15; protein kinase, mitogen activated kinase, 11, p38beta; reactive kinase; RK; SAPK/Erk/kinase 4; SAPK2; SAPK2A; SAPK2B; SAPK4; Serk4; Stress-activated protein kinase 2a; Stress-activated protein kinase 2b; Stress-activated protein kinase 4; stress-activated protein kinase-2; stress-activated protein kinase-2b; tRNA synthetase cofactor p38 | |
| A synthesized peptide derived from human p38 MAPK (150-250AA). | |
| Primary | |
| -20°C | |
| MAPK14 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title