missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 MAPK Mouse anti-Human, Porcine, Clone: 1A1C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66234-1-IG-150UL
This item is not returnable.
View return policy
Description
The p38 MAPK cascade regulates a variety of cellular responses to stress, inflammation, and other signals. p38 MAPK is relatively inactive in the nonphosphorylated form and becomes rapidly activated by dual phosphorylation of a Thr-Gly-Tyr motifs. There are four isoforms of p38 MAPK, which differ in their tissue expression and affinity for upstream activators and downstream effectors. When cells are exposed to tumor necrosis factor, interleukin-1, heat shock, or other activating stimuli, activation of MAPK kinase-3 occurs by phosphorylation. Activated MAPK kinase-3/6 phosphorylate each residue of Thr180 and Tyr182 in p38 MAPK. Phospho-p38 MAPK activates ATF-2, CHOP-1, MEF-2 and other transcription factors through phosphorylation.Specifications
| p38 MAPK | |
| Monoclonal | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16539 | |
| MAPK14 | |
| p38 MAPK Fusion Protein Ag5797 | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1A1C2 | |
| Unconjugated | |
| MAPK14 | |
| CSAID binding protein, CSBP, CSBP1, CSBP2, CSPB1, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, MAPK14, MAX interacting protein 2, Mxi2, p38, p38 MAPK, p38ALPHA, PRKM14, PRKM15, RK, SAPK2A | |
| Mouse | |
| Protein A | |
| RUO | |
| 100156630, 1432 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction