missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ p38 MAPK gamma Polyclonal Antibody
GREENER_CHOICE

Product Code. 16397874
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16397874 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16397874 Supplier Invitrogen™ Supplier No. PA580792

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ERK3/MAPK12/P38-gamma.

MAPK12 (p38 gamma) is a serine/threonine kinase involved in many different cellular processes. It is activated during hypoxia and gamma-irradiation-induced G(2) arrest. Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. MAPK12 functions as a signal transducer during differentiation of myoblasts to myotubes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p38 MAPK gamma
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Mapk12
Gene Accession No. O08911, P53778, Q63538
Gene Alias AW123708; ERK3; ERK5; erk6; ERK-6; etID309866.18; Extracellular signal-regulated kinase 6; fa05c12; LOW QUALITY PROTEIN: mitogen-activated protein kinase 12; mitogen-activated protein kinase 12; MAP kinase 12; MAP kinase p38 gamma; MAPK 12; mapk12; mapk12a; mitogen activated protein kinase 12; mitogen-activated protein kinase 12; mitogen-activated protein kinase 12a; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 gamma; OTTHUMP00000196671; P38gamma; p38-gamma; Prkm12; SAP kinase-3; sapk3; SAPK-3; stress activated protein kinase 3; stress-activated protein kinase 3; stress-activated protein kinase-3; wu:fa05c12; zgc:101695
Gene Symbols Mapk12
Host Species Rabbit
Immunogen Recombinant Human ERK3/MAPK12/P38-gamma Protein (Met1-Leu367).
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29857, 60352, 6300
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.