missing translation for 'onlineSavingsMsg'
Learn More

p38α inhibitor 1, MedChemExpress

Product Code. p-200097692
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491087 50 mg 50mg
16484828 10 mg 10mg
16477577 5 mg 5mg
16435626 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491087 Supplier Medchem Express Supplier No. HY114423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress p38α inhibitor 1 is a p38α inhibitor extracted from patent WO 2008076265 A1.

p38α inhibitor 1 is a p38α inhibitor extracted from patent WO 2008076265 A1.

Specifications

Chemical Name or Material p38α inhibitor 1
CAS 1034189-82-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H26F2N4O2
Quantity 50 mg
Solubility Information DMSO : ≥ 125 mg/mL (300.15 mM)
SMILES O=C(C1=CC2=C(C=C1OC3=CC=C(F)C=C3F)C=NN2CC(C)C)NCCN(C)C
Molecular Weight (g/mol) 416.46
Formula Weight 416.46
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.