missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 gamma/SAPK3 Antibody (OTI10E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47852
This item is not returnable.
View return policy
Description
p38 gamma/SAPK3 Monoclonal specifically detects p38 gamma/SAPK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| p38 gamma/SAPK3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| MAPK12 | |
| Full-length protein expressed in 293T cell transfected with human MAPK12 expression vector | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens mitogen-activated protein kinase 12 (MAPK12). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI10E1 | |
| Unconjugated | |
| EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3 | |
| Mouse | |
| 42 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6300 | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction