missing translation for 'onlineSavingsMsg'
Learn More

p38 gamma/SAPK3 Antibody (OTI10E1), DyLight 594, Novus Biologicals™

Product Code. 18428154 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428154 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428154 Supplier Novus Biologicals Supplier No. NBP273189DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p38 gamma/SAPK3 Monoclonal antibody specifically detects p38 gamma/SAPK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen p38 gamma/SAPK3
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI10E1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human p38 gamma/SAPK3 expression vector.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6300
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.