missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 gamma/SAPK3 Antibody (OTI10E1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73189CL1
This item is not returnable.
View return policy
Description
p38 gamma/SAPK3 Monoclonal antibody specifically detects p38 gamma/SAPK3 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| p38 gamma/SAPK3 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3 | |
| Full-length protein expressed in 293T cell transfected with human MAPK12 expression vector | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6300 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI10E1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction