missing translation for 'onlineSavingsMsg'
Learn More

p38 beta/MAPK11 Antibody (1F9), Novus Biologicals™

Product Code. 18369458 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369458 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369458 Supplier Novus Biologicals Supplier No. H00005600M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p38 beta/MAPK11 Monoclonal antibody specifically detects p38 beta/MAPK11 in Human samples. It is validated for Western Blot, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen p38 beta/MAPK11
Applications Western Blot, ELISA, KnockDown
Classification Monoclonal
Clone 1F9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH27933
Gene Alias EC 2.7.11, EC 2.7.11.24, MAP kinase 11, MAP kinase p38 beta, MAPK 11, mitogen-activated protein kinase 11, Mitogen-activated protein kinase p38 beta, mitogen-activated protein kinase p38-2, p38b, p38Beta, P38BETA2, PRKM11, SAPK2B, SAPK2p38-2P38B, Stress-activated protein kinase 2, stress-activated protein kinase-2, stress-activated protein kinase-2b
Host Species Mouse
Immunogen MAPK11 (AAH27933, 255 a.a. ~ 364 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ARTYIQSLPPMPQKDLSSIFRGANPLAIDLLGRMLVLDSDQRVSAAEALAHAYFSQYHDPEDEPEAEPYDESVEAKERTLEEWKELTYQEVLSFKPPEPPKPPGSLEIEQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5600
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.