Learn More
Description
PA1-848 detects p300 from human cells. PA1-848 has been successfully used in Western blot and ChIP procedures. By Western blot, this antibody detects an ~265 kDa protein representing p300 from HeLa cell lysate. The PA1-848 immunogen is a synthetic peptide corresponding to residues G(139) T S G P N Q G P T Q S T(151) C in the nuclear factor binding domain of human p300. The PA1-848 immunizing peptide (Cat. # PEP-053) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | p300 |
| Applications | ChIP Assay, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | EP300 |
| Gene Accession No. | Q09472 |
| Gene Alias | A430090G16; A730011L11; E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; Ep300; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; P300; p300 HAT; Protein propionyltransferase p300; RSTS2 |
| Gene Symbols | EP300 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.