missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p300 Monoclonal Antibody (RW105)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA116622
This item is not returnable.
View return policy
Description
Suggested positive control: HeLa nuclear extract.
EP300 encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. p300 is related by sequence to CPB (CREB-binding protein [CREB: cyclic-AMP responsive element binding protein]), and like CPB can stimulate transcription through activation of CREB. This EP300 activity is specifically inhibited by the adenovirus oncoprotein E1A. EP300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF.
Specifications
| p300 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| B2RWS6, Q09472 | |
| EP300 | |
| Fusion protein containing residues 1572-2371 of human KAT3B/p300. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Mustelid | |
| Antibody | |
| IgG1 κ |
| ChIP Assay, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| RW105 | |
| Unconjugated | |
| EP300 | |
| A430090G16; A730011L11; E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; Ep300; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; P300; p300 HAT; Protein propionyltransferase p300; RSTS2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 170915, 2033, 328572 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction