Learn More
Honeywell™ Freedom Half-Mask Air-Purifying Respirator (APR)
The Freedom Half-Mask APR is designed for occasional exposure to gases, vapours, and particulates. Limited use and maintenance-free, it is supplied pre-assembled with cartridges for elimination of assembly mistakes, replacement parts, and decontamination concerns.
Brand: Honeywell™ 1001597
Code : NEW
Additional Details : Weight : 0.80000kg
Alert:
To be disposed of when filters are saturated.
Description
- Product Type: Air-Purifying Respirator (APR)
- Mask Type: Half Mask
- Standards: EN405
- Cartridge Type: Dual Cartridge, Pre-Assembled
- Fastenings: One-Piece Sliding Suspension System
- Hazards: Gases, Organic Vapors (Temp. >65°C), Inorganic Vapors, Acids, Ammoniac and Derived Aminates (Class 1); Solid and Liquid Particulates (Class 3)
- For Use With (Equipment): Prefilter Pads (11847171, 12682743) and Pad Retaining Covers (12662653)
- For Use With (Applications): Chemical Industry, Construction Industry, Food Services, Manufacturing Industry, Medical Industry, Pharmaceutical Industry
Specifications
P3 | |
Green | |
Reusable | |
Prefilter Pads [11847171 (P1), 12682743 (P2)] and Pad Retaining Covers (12662653) | |
1 Ea. | |
Non-sterile | |
One-Piece Sliding Suspension System |
EN405 | |
Cup/Molded | |
General Purpose Particulate Respirator | |
P3 | |
Gases, Organic Vapors (Temp. >65°C), Inorganic Vapors, Acids, Ammoniac and Derived Aminates (Class 1); Solid and Liquid Particulates (Class 3) | |
Half-Mask |