missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ P3 Primary Cell 4D-Nucleofector™ X Kit S

Product Code. 12618553
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12618553 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12618553 Supplier Lonza™ Supplier No. V4XP3032
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For efficient transfection of specific primary cells, e.g., human T cells, CD34 or rat neurons, in the 4D-NucleofectorTM X Unit (20 μL format).

  • High transfection efficiencies for primary cells.
  • Cells maintain functionality post transfection.
  • Conditions are transferable between 4D-Nucleofector™ System, 96-well Shuttle™ System and 384-well Nucleofector™ System.
  • Applications:
    • Transfection of cell numbers in the range of 5 x 104 to 1 x 106 cells.
    • Medium throughput applications.
    • Optimization of transfection conditions, e.g., substrate titrations.
  • This kit has been determined to be optimal for transfecting following cell types:
    • B cell (human)
    • CD34+ cell (human)
    • Chondrocyte (human)
    • DC (human)
    • DC (mouse), mature
    • ES (human), H9
    • ES (mouse)
    • Hepatocyte (human)
    • HMEC (human)
    • Keratinocyte (human)
    • Macrophage (human)
    • Monocyte (human)
    • Neuron (rat), brain Hi/Cx
    • NHBE (human)
    • RPE (human)
    • T cell (human), stim.
    • T cell (mouse), unstim.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Vector pmaxGFP™
For Use With (Equipment) 4D-Nucleofector™ X Unit (20 μL format)
For Use With (Application) Efficient transfection of specific primary cells, e.g., human T cells, CD34 or rat neurons.
Includes P3 Primary Cell Nucleofector™ Solution, Supplement 1, pmaxGFP™ Vector (1 μg/μL in 10 mM Tris pH 8.0), 16-well Nucleocuvette™ Strips
No. of Reactions 32
pH 7.1 to 7.3
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.