missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab P3 Oxonia Active
Description
- P3-oxonia active, a peroxyacetic acid is effective against a broad spectrum of microorganisms.
- Effective at ambient temperatures, P3-oxonia active allows energy and time savings, avoiding waiting times associated with heating and cooling in thermal sanitisation.
Specifications
Specifications
| Product Type | Disinfectant |
| Form | Liquid |
| pH | 3.0 to 3.4 |
| Sterility | Non-sterile |
| Composition | Peroxyacetic Acid |
| Volume (Metric) | 22 kg |
Safety and Handling
Product Identifier
- P3-Oxonia Active
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H272-May intensify fire; oxidiser.
- H280-Contains gas under pressure; may explode if heated.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: hydrogen peroxide, acetic acid, peracetic acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?