missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA120697
This item is not returnable.
View return policy
Description
PA1-20697 detects P2Y8 in Human samples.
P2Y8 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. The gene codifying for this protein is moderately expressed in undifferentiated HL60 cells, and is located at both chromosomes X and Y.
Specifications
| P2Y8 | |
| Polyclonal | |
| Unconjugated | |
| P2RY8 | |
| G-protein coupled purinergic receptor P2Y8; MGC50878; P2RY8; P2Y purinoceptor 8; P2Y8; purinergic receptor P2Y, G-protein coupled, 8; purinergic receptor P2Y8; RP11-261P4.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286530 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q86VZ1 | |
| P2RY8 | |
| Synthetic 20 amino acid peptide from C-terminus of human P2RY8 / P2Y8. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction