missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y4/P2RY4 Antibody (3B4H6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | P2Y4/P2RY4 |
|---|---|
| Clone | 3B4H6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:200, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249771
|
Novus Biologicals
NBP2-61760 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637468
|
Novus Biologicals
NBP2-61760-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2Y4/P2RY4 Monoclonal specifically detects P2Y4/P2RY4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| P2Y4/P2RY4 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| GPCR | |
| 5030 | |
| Purified recombinant fragment of human P2RY4 (AA: extra mix) expressed in E. Coli. | |
| Primary | |
| 41 kDa |
| 3B4H6 | |
| Western Blot 1:500-1:200, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| P2P, P2Y4P2Y purinoceptor 4, pyrimidinergic receptor P2Y, G-protein coupled, 4, UNRNRUUridine nucleotide receptor | |
| P2RY4 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title