missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y2 Recombinant Rabbit Monoclonal Antibody (HL2704)

P2Y2 Recombinant Rabbit Monoclonal Antibody (HL2704)
Brand: Invitrogen MA556826
This item is not returnable.
View return policy
Description
P2Y2 Recombinant Monoclonal Antibody for WB, ICC/IF
Specifications
| P2Y2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P41231 | |
| P2RY2 | |
| Recombinant fragment of human P2Y2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2704 | |
| Unconjugated | |
| P2RY2 | |
| ATP receptor; HP2U; P2RU1; P2ry2; P2U; P2U nucleotide receptor; P2U purinoceptor 1; P2U receptor 1; P2U1; P2UR; P2Y ATP receptor 2; P2Y purinoceptor 2; P2Y2; P2Y2R; P2Y-like receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 2; purinergic receptor P2Y, G-protein coupled, 2; purinergic receptor P2Y2; purinoceptor P2Y2; R12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5029 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction