missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15060
This item is not returnable.
View return policy
Description
P2Y2 Polyclonal specifically detects P2Y2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| P2Y2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| ATP receptor, HP2U, MGC20088, MGC40010, P2RU1, P2U nucleotide receptor, P2U purinoceptor 1, P2U1, P2UP2UR, P2Y purinoceptor 2, P2Y2P2Y2R, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 2, purinergic receptor P2Y2, purinoceptor P2Y2 | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 5029 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunoprecipitation | |
| 0.82mg/mL | |
| Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500 | |
| P41231 | |
| P2RY2 | |
| Recombinant protein encompassing a sequence within the center region of human P2Y2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction