missing translation for 'onlineSavingsMsg'
Learn More

P2Y2 Antibody, Novus Biologicals™

Product Code. 18384705 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384705 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384705 Supplier Novus Biologicals Supplier No. NBP312229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

P2Y2 Polyclonal antibody specifically detects P2Y2 in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias ATP receptor, HP2U, MGC20088, MGC40010, P2RU1, P2U nucleotide receptor, P2U purinoceptor 1, P2U1, P2UP2UR, P2Y purinoceptor 2, P2Y2P2Y2R, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 2, purinergic receptor P2Y2, purinoceptor P2Y2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 360-410 on rat P2Y2 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR, Lipid and Metabolism, Myeloid derived Suppressor Cell, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5029
Target Species Human, Mouse, Rat, Feline
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.