missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
P2Y2 Polyclonal specifically detects P2Y2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | P2Y2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | ATP receptor, HP2U, MGC20088, MGC40010, P2RU1, P2U nucleotide receptor, P2U purinoceptor 1, P2U1, P2UP2UR, P2Y purinoceptor 2, P2Y2P2Y2R, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 2, purinergic receptor P2Y2, purinoceptor P2Y2 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of P2Y2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?