missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y14/GPR105 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86743
This item is not returnable.
View return policy
Description
P2Y14/GPR105 Polyclonal specifically detects P2Y14/GPR105 in Human samples. It is validated for Western Blot.
Specifications
| P2Y14/GPR105 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| G protein coupled receptor for UDP-glucose, GPR105, G-protein coupled receptor 105, KIAA0001G protein-coupled receptor 105, P2Y purinoceptor 14, P2Y(14) receptor, P2Y14, P2Y14 receptor, purinergic receptor P2Y, G-protein coupled, 14, UDP-glucose receptor | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of P2Y14/GPR105 | |
| 100 μL | |
| GPCR | |
| 9934 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction