missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y14/GPR105 Antibody (5A3F1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £438.00
Specifications
| Antigen | P2Y14/GPR105 |
|---|---|
| Clone | 5A3F1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18205521
|
Novus Biologicals
NBP2-61753 |
0.1 mL |
£438.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641969
|
Novus Biologicals
NBP2-61753-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2Y14/GPR105 Monoclonal specifically detects P2Y14/GPR105 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| P2Y14/GPR105 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| GPCR | |
| 9934 | |
| Purified recombinant fragment of human P2RY14 (AA: extra mix) expressed in E. Coli. | |
| Primary | |
| 39 kDa |
| 5A3F1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| G protein coupled receptor for UDP-glucose, GPR105, G-protein coupled receptor 105, KIAA0001G protein-coupled receptor 105, P2Y purinoceptor 14, P2Y(14) receptor, P2Y14, P2Y14 receptor, purinergic receptor P2Y, G-protein coupled, 14, UDP-glucose receptor | |
| P2RY14 | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title