missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P2Y13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534083
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset, Bovine, Bat (94%) Dog, Pig (81%).
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP.
Specifications
| P2Y13 | |
| Polyclonal | |
| Unconjugated | |
| P2RY13 | |
| 2010001L06Rik; FKSG77; G protein-coupled receptor 68; G protein-coupled receptor 86; GPCR1; GPR86; GPR94; G-protein coupled receptor 86; G-protein coupled receptor 94; P2ry13; P2Y purinoceptor 13; P2Y13; P2yr13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13; purinergic receptor P2Y13; SP174; Vpurinergic receptor P2Y, G-protein coupled 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53829 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9BPV8 | |
| P2RY13 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human P2RY13 / P2Y13. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction