missing translation for 'onlineSavingsMsg'
Learn More

P2Y13/P2RY13/GPR86 Antibody (6G12E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-52526-0.025mg

 View more versions of this product

Product Code. 18696108

  • £173.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

P2Y13/P2RY13/GPR86 Monoclonal specifically detects P2Y13/P2RY13/GPR86 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P2Y13/P2RY13/GPR86
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
G protein-coupled receptor 86, GPR86GPCR1, GPR94, G-protein coupled receptor 86, G-protein coupled receptor 94, P2Y purinoceptor 13, P2Y13FKSG77, purinergic receptor P2Y, G-protein coupled, 13, SP174
Mouse
40.8 kDa
0.025 mg
GPCR
53829
Human
Purified
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunohistochemistry (Paraffin)
6G12E10
Unconjugated
PBS with 0.05% Sodium Azide
P2RY13
Purified recombinant fragment of human P2RY13 (AA: 1-49) expresed in E. Coli
Protein A or G purified
RUO
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.