missing translation for 'onlineSavingsMsg'
Learn More

P2Y12/P2RY12 Antibody (1C2A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£184.00 - £469.00

Specifications

Antigen P2Y12/P2RY12
Clone 1C2A9
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18014786
View Documents
Novus Biologicals
NBP2-61749
0.1 mL
£469.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18686768
View Documents
Novus Biologicals
NBP2-61749-0.025ml
0.025 mL
£184.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

P2Y12/P2RY12 Monoclonal specifically detects P2Y12/P2RY12 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

P2Y12/P2RY12
1 mg/mL
Western Blot, Flow Cytometry, ELISA
Unconjugated
Mouse
ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999
P2RY12
IgG1
Protein G purified
1C2A9
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Monoclonal
Purified
GPCR
64805
Purified recombinant fragment of human P2RY12 (AA: extra mix) expressed in E. Coli.
Primary
39.4 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.