missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y11/P2RY11 Antibody (505214) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9305JF669
This item is not returnable.
View return policy
Description
P2Y11/P2RY11 Monoclonal antibody specifically detects P2Y11/P2RY11 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| P2Y11/P2RY11 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11 | |
| NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11, Accession # Q96G91 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 505214 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| 5032 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction