missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P2Y11 Monoclonal Antibody (505214)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA562014

Product Code. 30533835

  • £324.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

P2Y11 belongs to the family of G protein coupled receptors, and is coupled to the stimulation of the phosphoinositide and adenylyl cyclase pathways and behaves as a selective purinoceptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P2Y11
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q96G91
P2RY11
NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry
505214
Unconjugated
P2RY11
P2RY11; P2Y purinoceptor 11; P2Y11; purinergic receptor P2Y, G-protein coupled, 11; purinergic receptor P2Y11
Mouse
Protein A/G
RUO
5032
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.