missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y1/P2RY1 Antibody (4G5B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61671
This item is not returnable.
View return policy
Description
P2Y1/P2RY1 Monoclonal specifically detects P2Y1/P2RY1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| P2Y1/P2RY1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| ATP receptor, P2 purinoceptor subtype Y1, P2Y purinoceptor 1, P2Y1platelet ADP receptor, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 1 | |
| Mouse | |
| 42 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4G5B7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| P2RY1 | |
| Purified recombinant fragment of human P2RY1 (AA: extra mix) expressed in E. Coli. | |
| Protein G purified | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| 5028 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction