missing translation for 'onlineSavingsMsg'
Learn More

P2Y1/P2RY1 Antibody (4C2), Novus Biologicals™

Product Code. 18370449 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370449 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370449 Supplier Novus Biologicals Supplier No. H00005028M01

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

P2Y1/P2RY1 Monoclonal antibody specifically detects P2Y1/P2RY1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y1/P2RY1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4C2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002554
Gene Alias ATP receptor, P2 purinoceptor subtype Y1, P2Y purinoceptor 1, P2Y1platelet ADP receptor, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 1
Host Species Mouse
Immunogen P2RY1 (NP_002554, 1 a.a. ~ 52 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTEVLWPAVPNGTDAAFLAGPGSSWGNSTVASTAAVSSSFKCALTKTGFQFY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 5028
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.