missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7/P2RX7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-20180
This item is not returnable.
View return policy
Description
P2X7/P2RX7 Polyclonal specifically detects P2X7/P2RX7 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| P2X7/P2RX7 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 5027 | |
| Human, Mouse, Rat, Monkey | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
| Q9Z1M0 | |
| P2RX7 | |
| A synthetic peptide from mouse P2X7 conjugated to blue carrier protein was used as the antigen. | |
| 0.1 mL | |
| Primary | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size (0.025ml) is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction