missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7/P2RX7 Antibody (CL11508), Novus Biologicals™
Mouse Monoclonal Antibody
£317.00 - £511.00
Specifications
| Antigen | P2X7/P2RX7 |
|---|---|
| Clone | CL11508 |
| Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:500- 1:1000, Immunohistochemistry-Paraffin 1:500- 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625924
|
Novus Biologicals
NBP3-21196-100ul |
100 μg |
£511.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658594
|
Novus Biologicals
NBP3-21196-25ul |
25 μg |
£317.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| P2X7/P2RX7 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:500- 1:1000, Immunohistochemistry-Paraffin 1:500- 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q99572. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL11508 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Immunology, Signal Transduction | |
| PBS, pH 7.2, 40% glycerol | |
| 5027 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title