missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7/P2RX7 Antibody (CL11508) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21196AF594
This item is not returnable.
View return policy
Description
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| P2X7/P2RX7 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| This antibody was developed using a recombinant protein derived from Q99572, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 5027 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL11508 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction