missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7/P2RX7 Antibody (545205), Unconjugated, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB97901-SP
This item is not returnable.
View return policy
Description
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| P2X7/P2RX7 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 545205 | |
| Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| E. coli-derived partial recombinant human P2X7/P2RX7 protein (amino acids 356-595) | |
| 25 μg | |
| Immunology, Signal Transduction | |
| 5027 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction