missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7/P2RX7 Antibody (545205), FITC, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB97901F
This item is not returnable.
View return policy
Description
P2X7/P2RX7 Monoclonal specifically detects P2X7/P2RX7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| P2X7/P2RX7 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| P2RX7 | |
| E. coli-derived partial recombinant human P2X7/P2RX7 protein (amino acids 356-595) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 545205 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction