missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530112
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mmgt2 mediates Mg(2+) transport and may provide regulated pathways for Mg(2+) transport in the Golgi and post-Golgi organelles of epithelium-derived cells. Magnesium is an essential metal, but few selective transporters have been identified. The two separate gene products, Mmgt1 and Mmgt2, comprise the family that we have termed membrane Mg(2+) transporters (MMgTs), and the proteins reside in the membrane and mediate Mg(2+) transport. The transport of Mg(2+) is saturable and studies have identified that Mmgt2 fusion proteins reside in the Golgi complex and post-Golgi vesicles, including the early endosomes in COS-7 cells transfected with the respective tagged constructs. Among related pathways for Mmgt2 are transport and binding events, in particular, transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds.
Specifications
| P2X5 | |
| Polyclonal | |
| Unconjugated | |
| P2RX5 | |
| AI852046; ATP receptor; ATP receptor subunit; ionotropic ATP receptor P2X5; LRH-1; lymphoid-restricted histocompatibility antigen-1; membrane magnesium transporter 2; membrane magnesium transporter 2 {ECO:0000250; MGC47755; Mmgt2; mmgt2 {ECO:0000250; P2 purinergic receptor subtype; P2RX5; P2X purinoceptor 5; P2X5; P2X5 purinoceptor; P2x5 receptor; P2X5R; Purinergic receptor; purinergic receptor P2X 5; purinergic receptor P2X, ligand gated ion channel, 5; purinergic receptor P2X, ligand-gated ion channel, 5; purinergic receptor P2X5-1; purinergic receptor P2X5-2; RGD1311260; UniProtKB:Q8R3L0} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.99 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q93086 | |
| P2RX5 | |
| Recombinant fragment corresponding to a region within amino acids 1 and 228 of Human P2X5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur