missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ P2X4 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
A blocking peptide from P2X4. Source: Synthetic Amino Acid Sequence: (Accession #: NP_035156.2) C-ETDSVVSSVTTKAK The P2X4 Blocking Peptide is derived from Synthetic. The P2X4 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | Antibody Competition |
| Formulation | Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt. |
| Gene ID (Entrez) | 5025 |
| Name | P2RX4 Peptide |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -20 C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Gene Alias | ATP receptor, ATP-gated cation channel protein, P2RX4 |
| Gene Symbol | P2RX4 |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction