missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X2/P2RX2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38379-100ul
This item is not returnable.
View return policy
Description
P2X2/P2RX2 Polyclonal antibody specifically detects P2X2/P2RX2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| P2X2/P2RX2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ATP receptor, MGC129601, P2X purinoceptor 2, P2X2P2X Receptor, subunit 2, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-320 of human P2X2/P2RX2 (NP_733782.1).,, Sequence:, VFIVQKSYQESETGPESSIITKVKGITTSEHKVWDVEEYVKPPEGGSVFSIITRVEATHSQTQGTCPESIRVHNATCLSDADCVAGELDMLGNGLRTGRCVPYYQGPSKTCEVFGWCPVEDGASVSQFLGTMAPNFTILIKNSIHYPKFHFSKGNIADRTDGYLKRCTFHEASDLYCPIFKLGFIVEKAGESFTELAHKGGVIGVIINWDCDLDLPASECNPKYSFRRLDPKHVPASSGYNFRFAKYYKINGTTTRTLIKA | |
| 100 μL | |
| Signal Transduction | |
| 22953 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction